1-(Carbamoylmethyl)pyridinium chloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-(2-Amino-2-oxoethyl)pyridin-1-ium chloride

A0829-25G A0829-5G
TCIAA0829-25GEA 82 EUR
TCIAA0829-25G TCIAA0829-5G
1-(Carbamoylmethyl)pyridinium chloride ≥98.0% (by HPLC, titration analysis)
1-(Carbamoylmethyl)pyridinium chloride
Formula: C₇H₉ClN₂O
MW: 172,61 g/mol
Número MDL: MFCD00032002
Núm. CAS: 41220-29-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR