DELTA4-Androstene-3,17-dione ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
A0845-1G
A0845-5G
TCIAA0845-1GEA
26
EUR
TCIAA0845-1G
TCIAA0845-5G
DELTA4-Androstene-3,17-dione ≥98.0% (por GC)
DELTA4-Androstene-3,17-dione
Formula:
C₁₉H₂₆O₂ MW: 286,42 g/mol |
Número MDL:
MFCD00003615 Núm. CAS: 63-05-8 EINECS: 200-554-5 Índice Merck: 14,00638 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 171.0 to 175.0 deg-C |
Specific rotation [a]20/D | +188.0 to +193.0 deg (C=1, EtOH) |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...