N-Acetyl-DL-2-aminobutyric acid 97,0 Titrimetric
Proveedor: TCI
Sinónimos:
2-Acetamidobutyric Acid
A0853-25G
A0853-5G
TCIAA0853-25GEA
230
EUR
TCIAA0853-25G
TCIAA0853-5G
N-Acetyl-DL-2-aminobutyric acid 97,0 Titrimetric
N-Acetyl-DL-2-aminobutyric acid
Formula:
C₆H₁₁NO₃ MW: 145.16 g/mol |
Número MDL:
MFCD00020430 Núm. CAS: 7211-57-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...