Methyl-DL-2-aminobutyrate hydrochloride ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: H-Abu-OMe·HCl, DL-2-Aminobutyric Acid Methyl Ester Hydrochloride, Methyl 2-aminobutanoate hydrochloride

A0893-25G A0893-5G
TCIAA0893-25GEA 281 EUR
TCIAA0893-25G TCIAA0893-5G
Methyl-DL-2-aminobutyrate hydrochloride ≥98.0% (by total nitrogen basis)
Methyl-DL-2-aminobutyrate hydrochloride
Formula: C₅H₁₁NO₂
Número MDL: MFCD00058295
Núm. CAS: 7682-18-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR