Aminoacetonitrile sulphate ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Peligro

A0912-25G A0912-500G
TCIAA0912-25GEA 28 EUR
TCIAA0912-25G TCIAA0912-500G
Aminoacetonitrile sulphate ≥98.0% (by total nitrogen and titration analysis)
Aminoacetonitrile sulphate
Formula: C₄H₁₀N₄O₄S
Número MDL: MFCD00012851
Núm. CAS: 5466-22-8
EINECS: 226-772-0
UN: 3439
ADR: 6.1,III
Índice Merck: 14,00412

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR