2-Amino-2-oxazoline hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 4,5-Dihydrooxazol-2-amine hydrochloride

A0923-25G A0923-5G
TCIAA0923-25GEA 864 EUR
TCIAA0923-25G TCIAA0923-5G
2-Amino-2-oxazoline hydrochloride ≥98.0% (by titrimetric analysis)
2-Amino-2-oxazoline hydrochloride
Formula: C₃H₆N₂O
MW: 122,55 g/mol
Número MDL: MFCD00060178
Núm. CAS: 375855-07-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 114.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR