4-Azidobenzoic acid ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A0930.0005 A0930-25G A0930-5G
TCIAA0930.0005EA 53.2 EUR
TCIAA0930.0005 TCIAA0930-25G TCIAA0930-5G
4-Azidobenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Azidobenzoic acid
Formula: C₇H₅N₃O₂
Número MDL: MFCD00013987
Núm. CAS: 6427-66-3
EINECS: 229-198-9
UN: 1325
ADR: 4.1,III

Order Now

Resultados test especificaciones

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR