5-Amino-3-methyl-1-p-tolylpyrazole ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Methyl-1-(4-methylphenyl)-1H-pyrazol-5-amine

A0952-10G A0952-25G
TCIAA0952-10GEA 132 EUR
TCIAA0952-10G TCIAA0952-25G
5-Amino-3-methyl-1-p-tolylpyrazole ≥97.0% (by GC, titration analysis)
5-Amino-3-methyl-1-p-tolylpyrazole
Formula: C₁₁H₁₃N₃
MW: 187,25 g/mol
Número MDL: MFCD00020725
Núm. CAS: 62535-60-8
EINECS: 263-586-9

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 117.0 to 122.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR