Ethyl (2-amino-4-thiazolyl)acetate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
(2-Amino-4-thiazolyl)acetic Acid Ethyl Ester, Ethyl 2-(2-aminothiazol-4-yl)acetate, Ethyl 2-amino-4-thiazoleacetate
A0955-25G
A0955-500G
TCIAA0955-25GEA
25
EUR
TCIAA0955-25G
TCIAA0955-500G
Ethyl (2-amino-4-thiazolyl)acetate ≥98.0% (by GC, titration analysis)
Ethyl (2-amino-4-thiazolyl)acetate
Formula:
C₇H₁₀N₂O₂S MW: 186,23 g/mol |
Número MDL:
MFCD00005330 Núm. CAS: 53266-94-7 EINECS: 258-452-1 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...