D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

A1017-25G A1017-500G
TCIAA1017-25GEA 54 EUR
TCIAA1017-25G TCIAA1017-500G
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol ≥98.0% (by titrimetric analysis)
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Formula: C₉H₁₂N₂O₄
Número MDL: MFCD00078126
Núm. CAS: 716-61-0
EINECS: 211-938-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d22983013f9606dc212da0310df643d4f7332786&vid=d22983013f9606dc212da0310df643d4f7332786&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 162.0 to 167.0 deg-C
Specific rotation [a]20/D -29.0 to -32.0 deg (C=1, 6mol/L HCl)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR