1H-Pyrazolo[3,4-d]pyrimidin-4-amine ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

A1041-100MG A1041-1G A1041-5G
TCIAA1041-100MGEA 16 EUR
TCIAA1041-100MG TCIAA1041-1G TCIAA1041-5G
1H-Pyrazolo[3,4-d]pyrimidin-4-amine ≥97.0% (by HPLC, titration analysis)
1H-Pyrazolo[3,4-d]pyrimidin-4-amine
Formula: C₅H₅N₅
Número MDL: MFCD00005688
Núm. CAS: 2380-63-4
EINECS: 219-174-6
UN: 2811
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR