L-Azetidine-2-carboxylic acid ≥98.0% (by titrimetric analysis) antagonist of L-proline

Proveedor: TCI

A1043-1G A1043-100MG
TCIAA1043-1GEA 95 EUR
TCIAA1043-1G TCIAA1043-100MG
L-Azetidine-2-carboxylic acid ≥98.0% (by titrimetric analysis) antagonist of L-proline
L-Azetidine-2-carboxylic acid
Formula: C₄H₇NO₂
Número MDL: MFCD00005166
Núm. CAS: 2133-34-8
EINECS: 218-362-5
Índice Merck: 14,00908

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -120 to -125 deg (C=4, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR