2-Anthracenecarboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A1150-1G A1150-5G
TCIAA1150-1GEA 143 EUR
TCIAA1150-1G TCIAA1150-5G
2-Anthracenecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
Número MDL: MFCD00142865
Núm. CAS: 613-08-1

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR