5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

A1175-25G A1175-5G
TCIAA1175-25GEA 153 EUR
TCIAA1175-25G TCIAA1175-5G
5-Amino-1H-imidazole-4-carboxamide hydrochloride ≥98.0% (by HPLC, titration analysis)
5-Amino-1H-imidazole-4-carboxamide hydrochloride
Formula: C₄H₇ClN₄O
Número MDL: MFCD00012704
Núm. CAS: 72-40-2
EINECS: 200-778-3

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR