Aluminum glycinate ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate, Aluminum Glycinate
A1200-25G
A1200-500G
TCIAA1200-25GEA
17
EUR
TCIAA1200-25G
TCIAA1200-500G
Aluminum glycinate ≥97.0% (by titrimetric analysis)
Aluminum glycinate
Formula:
C₂H₆AlNO₄ MW: 135.05 g/mol |
Número MDL:
MFCD00053645 Núm. CAS: 13682-92-3 EINECS: 237-193-8 Índice Merck: 14,03183 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % (calcd.on anh.substance) |
Solubility in hot 1mol/L HCl | almost transparency |
Water | max. 14.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...