4-Amino-4'-nitrobiphenyl ≥96.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
A1251-1G
A1251-5G
TCIAA1251-1GEA
123
EUR
TCIAA1251-1G
TCIAA1251-5G
4-Amino-4'-nitrobiphenyl ≥96.0% (by titrimetric analysis)
4-Amino-4'-nitrobiphenyl
Formula:
C₁₂H₁₀N₂O₂ |
Número MDL:
MFCD00093527 Núm. CAS: 1211-40-1 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 200.0 to 205.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...