2-Aminociclohexanol cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1-Amino-2-hydroxycyclohexane, 2-Aminocyclohexanol (cis- and trans- mixture)
A1258-25G
A1258-5G
TCIAA1258-25GEA
256
EUR
TCIAA1258-25G
TCIAA1258-5G
2-Aminociclohexanol cis- and trans-mixture ≥98.0% (by GC, titration analysis)
2-Aminocyclohexanol
Formula:
C₆H₁₃NO MW: 115,18 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00191368 Núm. CAS: 6850-38-0 UN: 3259 ADR: 8,III |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % (total of isomer) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...