3-Amino-5,5-dimethyl-2-cyclohexen-1-one ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Amino-5,5-dimethylcyclohex-2-enone
A1300-25G
A1300-5G
TCIAA1300-25GEA
299
EUR
TCIAA1300-25G
TCIAA1300-5G
3-Amino-5,5-dimethyl-2-cyclohexen-1-one ≥98.0% (by GC, titration analysis)
3-Amino-5,5-dimethyl-2-cyclohexen-1-one
Formula:
C₈H₁₃NO MW: 139,20 g/mol |
Número MDL:
MFCD00045531 Núm. CAS: 873-95-0 |
Resultados test especificaciones
Appearance | White to Amber powder to crystalline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 164.0 to 168.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...