Ethyl-3-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl
A1310-10G
A1310-25G
TCIAA1310-10GEA
27
EUR
TCIAA1310-10G
TCIAA1310-25G
Ethyl-3-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
Ethyl-3-aminopropanoate hydrochloride
Formula:
C₅H₁₁NO₂·HCl MW: 153,6 g/mol |
Número MDL:
MFCD00012909 Núm. CAS: 4244-84-2 EINECS: 224-203-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 68.0 to 73.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...