2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride ≥93.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucose, 2-Acetamido-2-deoxy-α-D-glucopyranosyl chloride 3,4,6-triacetate, 2-Acetamido-2-deoxy-ɑ-D-glucopyranosyl Chloride 3,4,6-Triacetate, α-Acetochloro-D-glucosamine, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-ɑ-D-glucose, (2R,3S,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-chlorotetrahydro-2H-pyran-3,4-diyl diacetate, 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-glucopyranosyl chloride
A1416-1G
A1416-5G
TCIAA1416-1GEA
51
EUR
TCIAA1416-1G
TCIAA1416-5G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride ≥93.0% (by titrimetric analysis)
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl chloride
Formula:
C₁₄H₂₀ClNO₈ MW: 365,76 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00069776 Núm. CAS: 3068-34-6 EINECS: 221-325-6 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Argentmetric Titration) | min. 93.0 % |
Specific rotation [a]20/D | +110 to +125 deg (C=1, CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...