Sodium-4-acetylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 4'-Acetophenonesulphonic acid Sodium Salt, Sodium 4-acetylbenzenesulphonate, 4-Acetylbenzenesulphonic acid Sodium Salt

A1430-5G A1430-1G
TCIAA1430-5GEA 255 EUR
TCIAA1430-5G TCIAA1430-1G
Sodium-4-acetylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-acetylbenzenesulphonate
Formula: C₈H₈O₄S
MW: 222.19 g/mol
Número MDL: MFCD00007510
Núm. CAS: 61827-67-6
EINECS: 263-259-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR