N-Allyloxyphthalimide ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
A1447-25G
A1447-5G
TCIAA1447-25GEA
170
EUR
TCIAA1447-25G
TCIAA1447-5G
N-Allyloxyphthalimide ≥98.0% (por GC)
N-Allyloxyphthalimide
Formula:
C₁₁H₉NO₃ |
Número MDL:
MFCD00173307 Núm. CAS: 39020-79-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...