N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
N-[2-(4-Nitrophenyl)ethyl]acetamide, N-Acetyl-4-nitrophenethylamine
A1494-25G
A1494-5G
TCIAA1494-25GEA
329
EUR
TCIAA1494-25G
TCIAA1494-5G
N-Acetyl-2-(4-nitrophenyl)ethylamine ≥98.0% (por GC)
N-Acetyl-2-(4-nitrophenyl)ethylamine
Formula:
C₁₀H₁₂N₂O₃ MW: 208,22 g/mol |
Número MDL:
MFCD00024805 Núm. CAS: 6270-07-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...