1-Acetyladamantane ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
1-Adamantyl Methyl Ketone, Methyl 1-Adamantyl Ketone
A1571-25G
A1571-5G
TCIAA1571-25GEA
166
EUR
TCIAA1571-25G
TCIAA1571-5G
1-Acetyladamantane ≥98.0% (por GC)
1-Acetyladamantane
Formula:
C₁₂H₁₈O MW: 178,28 g/mol |
Número MDL:
MFCD00074739 Núm. CAS: 1660-04-4 EINECS: 216-761-9 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 56.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...