4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 1-[2-(4-Methoxyphenyl)ethyl]-4-aminopiperidine, 4-Amino-1-(4-methoxyphenethyl)piperidine, 1-(4-Methoxyphenethyl)-4-aminopiperidine

A1618-25G A1618-5G
TCIAA1618-25GEA 116 EUR
TCIAA1618-25G TCIAA1618-5G
4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine ≥98.0% (by HPLC, titration analysis)
4-Amino-1-[2-(4-methoxyphenyl)ethyl]piperidine
Formula: C₁₄H₂₂N₂O
MW: 234,34 g/mol
Número MDL: MFCD01631147
Núm. CAS: 85098-70-0
EINECS: 285-422-5

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR