1-Acetyl-4-(4-hydroxyphenyl)piperazine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(4-(4-Hydroxyphenyl)piperazin-1-yl)ethanone, 4-(4-Acetyl-1-piperazinyl)phenol
A1845-25G
A1845-250G
TCIAA1845-25GEA
22
EUR
TCIAA1845-25G
TCIAA1845-250G
1-Acetyl-4-(4-hydroxyphenyl)piperazine ≥98.0% (by GC, titration analysis)
1-Acetyl-4-(4-hydroxyphenyl)piperazine
Formula:
C₁₂H₁₆N₂O₂ MW: 220,27 g/mol |
Número MDL:
MFCD00044905 Núm. CAS: 67914-60-7 EINECS: 267-744-8 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 184.0 to 188.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...