5-(Acetoacetamido)-2-benzimidazolinone ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
5-Acetoacetlamino benzimdazolone
A1949-25G
A1949-250G
TCIAA1949-25GEA
38
EUR
TCIAA1949-25G
TCIAA1949-250G
5-(Acetoacetamido)-2-benzimidazolinone ≥98.0% (by HPLC, total nitrogen)
5-(Acetoacetamido)-2-benzimidazolinone
Formula:
C₁₁H₉N₃O₃ MW: 231,21 g/mol |
Número MDL:
MFCD00444508 Núm. CAS: 26576-46-5 EINECS: 247-820-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...