5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Amino-2-benzimidazolinone
A1950-25G
A1950-5G
TCIAA1950-25GEA
95
EUR
TCIAA1950-25G
TCIAA1950-5G
5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥98.0% (by HPLC)
5-Amino-1,3-dihydro-2H-benzimidazol-2-one
Formula:
C₇H₇N₃O MW: 149,1500 g/mol |
Número MDL:
MFCD00053555 Núm. CAS: 95-23-8 EINECS: 202-401-8 |
Resultados test especificaciones
Appearance | White to Amber to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...