Azithromycin dihydrate ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: Azithromycin hydrate, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-(((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-2-ethyl-3,4,10-trihydroxy-13-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one dihydrate

A2076-1G A2076-5G
TCIAA2076-1GEA 55 EUR
TCIAA2076-1G TCIAA2076-5G
Azithromycin dihydrate ≥98.0% (by total nitrogen basis)
Azithromycin dihydrate
Formula: C₃₈H₇₂N₂O₁₂·2H₂O
MW: 749.00 g/mol
Número MDL: MFCD01862248
Núm. CAS: 117772-70-0
Índice Merck: 14,00915

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -45.0 to -49.0 deg (C=2, EtOH) (calcd.on anh.substance)
Water 4.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR