5-(Aminosulphonyl)-2,3-dimethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2,3-Dimethoxy-5-sulphamoylbenzoic acid

A2126-25G A2126-5G
TCIAA2126-25GEA 87 EUR
TCIAA2126-25G TCIAA2126-5G
5-(Aminosulphonyl)-2,3-dimethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
5-(Aminosulphonyl)-2,3-dimethoxybenzoic acid
Formula: C₉H₁₁NO₆S
MW: 261,25 g/mol
Número MDL: MFCD01317561
Núm. CAS: 66644-80-2
EINECS: 266-433-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 203.0 to 209.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR