Artesunate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
A2191-25G
A2191-5G
TCIAA2191-25GEA
155
EUR
TCIAA2191-25G
TCIAA2191-5G
Artesunate ≥98.0% (by HPLC, titration analysis)
Artesunate
Formula:
C₁₉H₂₈O₈ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00866204 Núm. CAS: 88495-63-0 Índice Merck: 14,00818 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 132.0 to 137.0 deg-C |
Specific rotation [a]20/D | +4.5 to +6.5 deg (C=1, CH2Cl2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...