3-Amino-2-hydroxy-5-nitrobenzenesulphonic acid monohydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Amino-2-hydroxy-5-nitrobenzenesulphonic Acid Monohydrate

A2197-25G A2197-5G
TCIAA2197-25GEA 171 EUR
TCIAA2197-25G TCIAA2197-5G
3-Amino-2-hydroxy-5-nitrobenzenesulphonic acid monohydrate ≥98.0% (by HPLC, titration analysis)
3-Amino-2-hydroxy-5-nitrobenzenesulphonic acid monohydrate
Formula: C₆H₆N₂O₆S
MW: 234,18 g/mol
Número MDL: MFCD00035758
Núm. CAS: 96-67-3
EINECS: 202-523-1

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 6.5 to 8.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR