N-(3-Aminopropyl)-2-nitrobenzenesulphonamide hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: N-(2-Nitrobenzenesulphonyl)-1,3-propanediamine Hydrochloride, N-(o-Ns)-1,3-diaminopropane Hydrochloride, 1-Amino-3-(2-nitrobenzenesulphonamido)propane Hydrochloride

A2268-1G A2268-5G
TCIAA2268-1GEA 103 EUR
TCIAA2268-1G TCIAA2268-5G
N-(3-Aminopropyl)-2-nitrobenzenesulphonamide hydrochloride ≥98.0% (by titrimetric analysis)
N-(3-Aminopropyl)-2-nitrobenzenesulphonamide hydrochloride
Formula: C₉H₁₃N₃O₄S
MW: 295,74 g/mol
Número MDL: MFCD07366920
Núm. CAS: 863983-46-4

Order Now

Resultados test especificaciones

Appearance White to Light red to Green powder to crystalline
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR