Amantadina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-Adamantylamine, 1-Aminotricyclo[3.3.1.1(3,7)]decane, Adamantan-1-amine, 1-Adamantanamine, 1-Aminoadamantane

A2318-5G A2318-25G
TCIAA2318-5GEA 48 EUR
TCIAA2318-5G TCIAA2318-25G
Amantadina ≥98.0% (by GC, titration analysis)
Amantadine
Formula: C₁₀H₁₇N
MW: 151,25 g/mol
Número MDL: MFCD00074732
Núm. CAS: 768-94-5
EINECS: 212-201-2
UN: 3259
ADR: 8,II
Índice Merck: 14,00374
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ba7488f9b0bda3b9fc934db0d230350d0b7ac0&vid=22ba7488f9b0bda3b9fc934db0d230350d0b7ac0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR