Azelastine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)phthalazin-1(2H)-one hydrochloride, Azelastine HCl, 4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride

A2340-1G A2340-100MG
TCIAA2340-1GEA 167 EUR
TCIAA2340-1G TCIAA2340-100MG
Azelastine hydrochloride ≥98.0% (by HPLC, titration analysis)
Azelastine hydrochloride
Formula: C₂₂H₂₄ClN₃O
MW: 418,36 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00242783
Núm. CAS: 79307-93-0
Índice Merck: 14,00906

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 224.0 to 228.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR