5-Chloro-3-methylanthranilic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Amino-5-chloro-3-methylbenzoic acid

A2399-5G A2399-25G
TCIAA2399-5GEA 31 EUR
TCIAA2399-5G TCIAA2399-25G
5-Chloro-3-methylanthranilic acid ≥98.0% (by HPLC, titration analysis)
5-Chloro-3-methylanthranilic acid
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
Número MDL: MFCD02358895
Núm. CAS: 20776-67-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR