2-Amino-6-(trifluoromethoxy)benzothiazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole
A2423-5G
A2423-25G
TCIAA2423-5GEA
43
EUR
TCIAA2423-5G
TCIAA2423-25G
2-Amino-6-(trifluoromethoxy)benzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-6-(trifluoromethoxy)benzothiazole
Formula:
C₈H₅F₃N₂OS MW: 234,20 g/mol |
Número MDL:
MFCD00210213 Núm. CAS: 1744-22-5 UN: 2811 ADR: 6.1,II Índice Merck: 14,08223 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...