3-Amino-4-hydroxypyridine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Amino-4-pyridinol

A2432-1G A2432-5G
TCIAA2432-1GEA 23 EUR
TCIAA2432-1G TCIAA2432-5G
3-Amino-4-hydroxypyridine ≥98.0% (by GC, titration analysis)
3-Amino-4-hydroxypyridine
Formula: C₅H₆N₂O
MW: 110,12 g/mol
Número MDL: MFCD04114230
Núm. CAS: 6320-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7653560338524b63011a39586e555a9e9cdb22d&vid=28004ddc297b6e04e65e10c45ab966f615f93391&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Melting point 148.0 to 153.0 deg-C
Drying loss max. 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR