Azelnidipine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl Ester, 3-(1-Benzhydrylazetidin-3-yl) 5-isopropyl 2-amino-6-methyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, 3-[1-(Diphenylmethyl)-3-azetidinyl] 5-Isopropyl 2-Amino-1,4-dihydro-6-methyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

A2433-1G A2433-5G
TCIAA2433-1GEA 50 EUR
TCIAA2433-1G TCIAA2433-5G
Azelnidipine ≥98.0% (by HPLC, titration analysis)
Azelnidipine
Formula: C₃₃H₃₄N₄O₆
MW: 582,66 g/mol
Número MDL: MFCD00865803
Núm. CAS: 123524-52-7
Índice Merck: 14,00907

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR