N-(2-Aminoethyl)maleimide hydrochloride ≥93.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: N-(2-Aminoethyl)maleimide HCl, N-(2-Aminoethyl)maleimide Hydrochloride, 2-Maleimidoethylamine hydrochloride

A2436-1G A2436-200MG
TCIAA2436-1GEA 274 EUR
TCIAA2436-1G TCIAA2436-200MG
N-(2-Aminoethyl)maleimide hydrochloride ≥93.0% (by HPLC, total nitrogen)
N-(2-Aminoethyl)maleimide hydrochloride
Formula: C₆H₈N₂O₂
MW: 176,60 g/mol
Número MDL: MFCD09800491
Núm. CAS: 134272-64-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (with Total Nitrogen) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR