Azetidine hydrochloride ≥95.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: Azetidine HCl

A2451-5G A2451-1G
TCIAA2451-5GEA 144 EUR
TCIAA2451-5G TCIAA2451-1G
Azetidine hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Azetidine hydrochloride
Formula: C₃H₈ClN
MW: 93,56 g/mol
Número MDL: MFCD00191762
Núm. CAS: 36520-39-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR