(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid ≥95.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

A2470-5G A2470-1G
TCIAA2470-5GEA 87 EUR
TCIAA2470-5G TCIAA2470-1G
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid ≥95.0% (by HPLC, titration analysis)
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
Formula: C₈H₁₆N₂O₄
MW: 204,23 g/mol
Pto. de fusión: 210°C
Número MDL: MFCD00236843
Núm. CAS: 73259-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41ee45414b1b039ebb47edeabb7c499ca2276cf8&vid=41ee45414b1b039ebb47edeabb7c499ca2276cf8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D -4.0 to -6.0 deg (C=1、AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR