(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: Z-Dap-OH, Nα-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 3-Amino-N-(carbobenzoxy)-L-alanine, N(α)-Z-L-2,3-Diaminopropionic acid, (S)-Nalpha-Cbz-2,3-diaminopropionic Acid, Cbz-Dap-OH, Z-Dpr-OH, 3-Amino-N-Cbz-L-alanine

A2471-5G A2471-1G
TCIAA2471-5GEA 118 EUR
TCIAA2471-5G TCIAA2471-1G
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-3-Amino-2-(((benzyloxy)carbonyl)amino)propanoic acid
Formula: C₁₁H₁₄N₂O₄
MW: 238,5 g/mol
Pto. de fusión: 230°C
Número MDL: MFCD00237345
Núm. CAS: 35761-26-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9801cbcac1cf0bce8b5b34cbac049ed3a5f253fd&vid=9801cbcac1cf0bce8b5b34cbac049ed3a5f253fd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -11.0 to -14.0 deg (C=1、1mol/L NaOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR