4-Acetamido-N-(2-aminophenyl)benzamide ≥98.0% (by HPLC)

Proveedor: TCI

A2501-10MG A2501-50MG
TCIAA2501-10MGEA 42 EUR
TCIAA2501-10MG TCIAA2501-50MG
4-Acetamido-N-(2-aminophenyl)benzamide ≥98.0% (by HPLC)
4-Acetamido-N-(2-aminophenyl)benzamide
Formula: C₁₅H₁₅N₃O₂
Temperatura de almacenaje: Nevera
Número MDL: MFCD00866266
Núm. CAS: 112522-64-2

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 15.0 to 16.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR