2-Amino-4-(trifluorometil)pirimidina 98,0 GC, T
Proveedor: TCI
|
Atención
|
A2522-1G
A2522-5G
TCIAA2522-1GEA
37
EUR
TCIAA2522-1G
TCIAA2522-5G
2-Amino-4-(trifluorometil)pirimidina 98,0 GC, T
2-Amino-4-(trifluorometil)pirimidina
Formula:
C₅H₄F₃N₃ MW: 163.1 g/mol |
Número MDL:
MFCD00828624 Núm. CAS: 16075-42-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...