2-Amino-4-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Amino-p-toluic acid, 4-Methylanthranilic acid, 4-Methylanthranil acid, 2-Amino-4-methylbenzoic Acid
A2538-1G
A2538-5G
TCIAA2538-1GEA
104
EUR
TCIAA2538-1G
TCIAA2538-5G
2-Amino-4-methylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-4-methylbenzoic acid
Formula:
C₈H₉NO₂ MW: 151,16 g/mol |
Número MDL:
MFCD00047853 Núm. CAS: 2305-36-4 EINECS: 218-976-3 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...