4-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A2543-1G A2543-5G
TCIAA2543-1GEA 36 EUR
TCIAA2543-1G TCIAA2543-5G
4-Amino-2-methoxypyridine ≥98.0% (by GC, titration analysis)
4-Amino-2-methoxypyridine
Formula: C₆H₈N₂O
Número MDL: MFCD06738657
Núm. CAS: 20265-39-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR