2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2-imino-4-thiazolidinone, 2-Amino-5-(4-(2-(5-ethylpyridin-2-yl)ethoxy)benzyl)thiazol-4(5H)-one

A2567-5G A2567-25G
TCIAA2567-5GEA 76 EUR
TCIAA2567-5G TCIAA2567-25G
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one ≥98.0% (by HPLC, titration analysis)
2-Amino-5-[4-[2-(5-ethyl-2-pyridyl)ethoxy]benzyl]thiazol-4(5H)-one
Formula: C₁₉H₂₁N₃O₂S
MW: 355,46 g/mol
Número MDL: MFCD07368627
Núm. CAS: 105355-26-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR