Amprolium hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
Proveedor: TCI
Sinónimos:
Amprolium HCl, 1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride, Amprol, 1-((4-Amino-2-propylpyrimidin-5-yl)methyl)-2-methylpyridin-1-ium chloride hydrochloride, Amprolium hydrochloride, 1-[(4-Amino-2-propyl-5-pyrimidinyl)methyl]-2-methylpyridinium Chloride Hydrochloride, Mepyrium, Thiacoccid
A2572-5G
A2572-25G
TCIAA2572-5GEA
16
EUR
TCIAA2572-5G
TCIAA2572-25G
Amprolium hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
Amprolium hydrochloride
Formula:
C₁₄H₁₉ClN₄ MW: 315,24 g/mol |
Número MDL:
MFCD00078831 Núm. CAS: 137-88-2 EINECS: 205-307-5 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 239.0 to 243.0 deg-C |
Suitability for protein-analysis | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...