(S)-Isopropyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (S)-Isopropyl 2-aminopropanoate hydrochloride, (S)-2-Aminopropionic acid Isopropyl ester hydrochloride, L-Alanine Isopropyl ester hydrochloride, H-Ala-OiPr·HCl, Isopropyl (S)-2-Aminopropionate hydrochloride

A2588-5G A2588-25G
TCIAA2588-5GEA 27 EUR
TCIAA2588-5G TCIAA2588-25G
(S)-Isopropyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(S)-Isopropyl-2-aminopropanoate hydrochloride
Formula: C₆H₁₃NO₂·HCl
MW: 167.6 g/mol
Número MDL: MFCD08059709
Núm. CAS: 62062-65-1

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +1.0 to +3.0 deg (C=2,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR